Learn More
Bochem™ Sharp Forceps
Bochem™ Sharp Forceps have a pointed end and are constructed of Stainless Steel.
Brand: Bochem™ 1015
This item is not returnable.
View return policy
Description
- Stainless steel
- Pointed tips
Specifications
Sharp | |
Stainless Steel (18/10) | |
200 mm |
Product Content Correction
Product Title
Bochem™ Sharp Forceps > Length: 200mm
Spot an opportunity for improvement?Share a Content Correction