missing translation for 'onlineSavingsMsg'
Learn More

Woodsafe™ Sharpsbin and Hazardous waste containers with double lid in natural color
GREENER_CHOICE

Woodsafe is a cost-effective solution for reducing the use of fossil based plastic by 80% and CO2 emissions by 66%, documented in in a full LCA. Produced with residue from the forest industry using green energy. Available in sizes from 0.5 L to 50 L. Single and double lid versions.

1074.61 DKK - 20329.00 DKK

Specifications

Certifications/Compliance EN ISO 23907-1:2019. UN3291, ADR Approved
Closure Type Temporary and permanent closure
Closure Color Brown
Description Environmental friendly Sharpsbin
Color Brown, Yellow
View More Specs

Alert:

Follow instructions for safe and correct use


Products 7
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
18737338
GREENER_CHOICE
View accessory products View Documents
Woodsafe™
1005120
0.5 L
1074.61 DKK
Pack of 50
Please to purchase this item. Need a web account? Register with us today!
18727338
GREENER_CHOICE
View accessory products View Documents
Woodsafe™
1020120
2.3 L
2174.85 DKK
Pack of 70
Please to purchase this item. Need a web account? Register with us today!
18106549
GREENER_CHOICE
View accessory products View Documents
Woodsafe™
1030120
3.3 L
1843.64 DKK
Pack of 50
Please to purchase this item. Need a web account? Register with us today!
18186539
GREENER_CHOICE
View accessory products View Documents
Woodsafe™
1060120
6 L
2087.27 DKK
Pack of 30
Please to purchase this item. Need a web account? Register with us today!
18126539
GREENER_CHOICE
View accessory products View Documents
Woodsafe™
1120120
12 L
1871.52 DKK
Pack of 20
Please to purchase this item. Need a web account? Register with us today!
18792815
GREENER_CHOICE
View accessory products View Documents
Woodsafe™
1250120
25 L
18033.18 DKK
Pack of 132
Please to purchase this item. Need a web account? Register with us today!
18772825
GREENER_CHOICE
View accessory products View Documents
Woodsafe™
1500120
50 L
20329.00 DKK
Pack of 96
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Woodsafe™ offer a sustainable clinical waste management solution produced with residue from the Swedish forest industry
  • Lowers the use of Fossil-based plastic by 80%!
  • Lowers the carbon footprint by more than 60%, proved in full LCA
  • Made from 100% Biocomposite
  • The lid can be securely permanently closed for transportation to the final disposal
  • My Green Labs ACT Label
  • Follows same process as yellow bins and can be used to transport the same substances
  • Natural color and smell from wood - No migration of substances
  • Compliant with UN3291
  • Tested according to ISO 23907-1:2019
  • UN ADR Approved
  • Biocomposite have the same properties as fossil-based plastic and can be used exactly the same way
  • NOTE: String between lids is for delivery purpose, remove/break before use!
  • Stop burning plastic
Specifications

Specifications

EN ISO 23907-1:2019. UN3291, ADR Approved
Brown
Brown, Yellow
Non-sterile
Biocomposite
Temporary and permanent closure
Environmental friendly Sharpsbin
Single-use
Autoclavable
Biohazard Waste Container (Benchtop)
Videos
SDS
Documents

Documents

Product Certifications

Lower climate impact and ACT label

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.