missing translation for 'onlineSavingsMsg'
Learn More

Woodsafe Sharpsbin
GREENER_CHOICE

A cost-effective solution for lowering the CO2 emissions. Eco-friendly sharps and hazardous waste containers, produced from renewable resources using green energy. Reducing CO2 emissions by up to 66%, documented in a full LCA. Available in sizes from 0.5 L to 50 L.

Brand:  Woodsafe 1120110

 View more versions of this product

Product Code. 18762825

  • 1590.05 DKK / Pack of 20

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

Follow instructions for safe and correct use

This item is not returnable. View return policy

Description

Description

  • Woodsafe™ offer a sustainable clinical waste management solution produced with residue from the forest industry.
  • Lowers the carbon footprint by more than 60%, proved in full LCA
  • Made from 100% Biocomposite
  • The lid can be temporarily closed to conceal the contents when not in use and securely permanently closed for transportation to the final disposal.
  • Natural colour and smell from wood - No migration of substances - EN - tested for toys and food
  • Compliant with UN3291
  • Tested according to ISO 23907-1:2019
  • UN ADR Approved
  • Biocomposite have the same properties as fossil-based plastic and can be used exactly the same way
  • NOTE: String between lids is for delivery purpose, remove/break before use!
  • Stop burning plastic
Specifications

Specifications

12 L
Permanent closure
Environmental friendly Sharpsbin
Brown, Yellow
Single-use
Yes
Non-sterile
Bin + Lid + Label
Cardboard
Biohazard Waste Container (Benchtop)
EN ISO 23907-1:2019. UN3291, ADR Approved
Brown
3.17 gal.
31.5 x 18.6 x 34 cm (12.4 x 7.32 x 13.39 in.)
20
31.5 x 18.6 x 34 cm
Autoclavable
Safe Disposal of sharps and potentially Infectious Waste or medical waste
Biocomposite
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.