Learn More
Fisherbrand™ Shell Vial Kit, Clear Glass, PE Plug
Recommended for HPLC usage.
135.00 DKK - 173.00 DKK
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
11555914
|
Fisherbrand™
11 14 0544 |
2.3 mL |
135.00 DKK
Pack of 100 |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||||
11516074
|
Fisherbrand™
15 14 0548 |
5.5 mL |
173.00 DKK
Pack of 100 |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||||
Description
- Shell vials and the appropriate plugs can also be supplied as a 2in1 KIT
- Handy and inexpensive vial/ closure combination for uncritical analyses
Specifications
Shell Vial | |
Clear | |
Glass | |
No |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Shell Vial Kit, Clear Glass, PE Plug