Learn More
Bohlender™ SICCO™ Mini Desiccator for Gas Filling Basic
Mini Desiccator for Gas Filling Basic made of polycarbonate, inlcludes desiccant.
Brand:  Bohlender™ V1849-06
Product Code. 17157704
  • 3145.00 DKK / Each
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Description
  • Made of polycarbonate
  • Includes desiccant and hose (5 m) with two quick couplings for gas filling
  • Stackable, with One Touch Door with circular seal
  • Robust construction
  • Transparent, shatterproof cabinet made of polycarbonate
  • Superb all-round visibility
  • Hinge door with wide opening angle
  • Lightweight, transportable
  • Two connections for gas filling including quick couplings with self-sealing valves and hose
  • Desiccant in a handy, resealable container included in delivery
  • Additional place requirement for coupling: 150 mm (6 in.)
Specifications
Specifications
Polycarbonate
1.6 gal.
8.7 in.
7 in.
8.4 in.
2 lb.
Silica Gel Jar (23 g)
6.2 L
221 mm
183 mm
214 mm
0.9 kg