Learn More
RETSCH Sieving Aids Rubber Balls
Brand:  RETSCH 32.354.0001
Product Code. 10156791
  • 2145.00 DKK / Pack of 5
Estimated Shipment: 04-07-2025
to see stock.
Explore more special offers
Specifications
Rubber Balls
20 mm
Horizontal Sieve Shaker AS 400 control
5/Pk.
Product Title
RETSCH Sieving Aids Rubber Balls > Diameter: 20mm