Learn More
Siglec-G Antibody (805903), PE/Cy5.5, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB7775PECY55
Product Code. 30032285
  • 4795.00 DKK / 0.10mL
Explore more special offers
This item is not returnable. View return policy
Description
Description
Siglec-G Monoclonal antibody specifically detects Siglec-G in Mouse samples. It is validated for Flow Cytometry
Specifications
Specifications
Siglec-G
Monoclonal
PE-Cyanine5.5
PBS
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
89790
Store at 4C in the dark. Do not freeze.
IgG2a
Flow Cytometry
805903
Flow Cytometry
PRO940;Sialic acid-binding Ig-like lectin 10;SIGLEC10;SIGLEC-10;SLG2
Chinese hamster ovary cell line CHO-derived recombinant mouse Siglec-G, Glu20-Gln525, Accession # NP_766488
0.1 mL
Primary
Mouse
Purified