missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Siglec-G Mouse anti-Mouse, BUV661, Clone: SH1, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 750295
4990.48 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Siglec-G Monoclonal antibody specifically detects Siglec-G in Mouse samples. It is validated for Flow Cytometry
Specifications
Siglec-G | |
Monoclonal | |
Brilliant Ultraviolet 661 | |
Siglecg; mSiglec-G; Siglec10; Siglec 10; CD330 | |
Mouse Extracellular Siglec-G Recombinant Protein | |
RUO | |
Mouse | |
IgG1 κ |
Flow Cytometry | |
SH1 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV661 under optimal conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Siglec-G Mouse anti-Mouse, BUV661, Clone: SH1, BD Biosciences™ > 50 μg; BUV661
Spot an opportunity for improvement?Share a Content Correction