Learn More
Thermo Scientific Chemicals Silica gel, wide pore, 150 angstroms
Description
Silica gel, wide pore is mainly used for the dehydrated purification of the industrial gases, the clearance of the organic acids and high polymers in the insulative oil, the purification and separation of the medicines. It is also used in column chromatography without applied pressure (gravity pulled).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Particle Size | -100/200 Mesh |
Pore Size | 150 Å |
Packaging | Bottle |
pH | 6.5 to 7.5 |
Quantity | 250 g |
Surface Area (Metric) | 350 to 400 m2/g |
Type | Silica Gel |
Material | Silica |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Q29K026 | Certificate of Analysis | 17/05/2024 | 042726.30,042726.18,042726.PG,042726.A7,042726.A1 |
B23L19 | Certificate of Analysis | 22/11/2021 | 042726.30,042726.18,042726.PG,042726.A7,042726.A1 |
G05L36 | Certificate of Analysis | 22/11/2021 | 042726.30,042726.18,042726.PG,042726.A7,042726.A1 |
E03S002 | Certificate of Analysis | 22/11/2021 | 042726.30,042726.18,042726.PG,042726.A7,042726.A1 |
W27F007 | Certificate of Analysis | 22/11/2021 | 042726.30,042726.18,042726.PG,042726.A7,042726.A1 |