Learn More
Silicon, plasma standard solution, Specpure™ Si 1000μg/mL, Thermo Scientific Chemicals Product Code.: 11338997
Silicon, plasma standard solution, Specpure™ Si 1000μg/mL, Thermo Scientific Chemicals
Product Code. 11338997
50 mL
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Product Code. 11338997
Brand: Thermo Scientific Alfa Aesar 038717.AD
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
MDL Number MFCD00085311
Specifications
Quantity 50 mL
Physical Form Liquid
Grade Specpure™
Chemical Name or Material Silicon plasma standard solution, Si 1000μg/mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1546374 Certificate TypeCertificate of Analysis Date07/04/2025 Catalog Number038717.AP,038717.AD,038717.AE
Lot Number1543951 Certificate TypeCertificate of Analysis Date17/03/2025 Catalog Number038717.AP,038717.AD,038717.AE
Lot Number1533783 Certificate TypeCertificate of Analysis Date14/03/2025 Catalog Number038717.AP,038717.AD,038717.AE
Lot Number1533791 Certificate TypeCertificate of Analysis Date13/03/2025 Catalog Number038717.AP,038717.AD,038717.AE
Lot Number1525850 Certificate TypeCertificate of Analysis Date13/03/2025 Catalog Number038717.AP,038717.AD,038717.AE
RUO – Research Use Only
Product Content Correction
Product Title
Silicon, plasma standard solution, Specpure™ Si 1000μg/mL, Thermo Scientific Chemicals >