Learn More
Thermo Scientific™ Silicone-free Viton™ Door Sealing for Thermo Scientific™ Heratherm™ Incubators and Ovens
For use with Thermo Scientific™ Heratherm™ Incubators and Ovens
Brand:  Thermo Scientific™ 50135869
 View more versions of this product
Product Code. 12615326
  • 6148.58 DKK / Each
Estimated Shipment: 17-06-2025
to see stock.
Explore more special offers
Specifications
Heratherm 400l Models
Door Seal
Videos
  • Thermo Scientific - Heratherm heating and drying ovens
    Thermo Scientific Heratherm heating and drying ovens are efficient, safe and easy to use.
    Video Player is loading.
    Current Time 0:00
    Duration 2:20
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 2:20
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Thermo Scientific Heratherm heating and drying ovens
  • Thermo Scientific - Heratherm heating and drying ovens
    Thermo Scientific Heratherm heating and drying ovens are efficient, safe and easy to use.
    Video Player is loading.
    Current Time 0:00
    Duration 2:20
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 2:20
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Thermo Scientific Heratherm heating and drying ovens
Product Title
Thermo Scientific™ Silicone-free Viton™ Door Sealing for Thermo Scientific™ Heratherm™ Incubators and Ovens > Silicone-free viton sealing for all Heratherm 400L models