Learn More
IKA™ H 100 Silicone Protective Cover
Use this protective cover with RCT basic magnetic stirrers. IKA™ H 100 Silicone Protective Covers are resistant to most acids, alkaline solutions and organic solvents.
Brand:  IKA™ 0003661000
162.04 DKK valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Product Code. 10189972
  • 186.00 DKK / Each
In Stock
Explore more special offers
Description
  • Silicone  protective cover
  • For RCT basic s.c. (3380000 + 3810000), included with the magnetic stirrer
  • Max. temperature 135°C
TRUSTED_SUSTAINABILITY
Specifications
Specifications
135°C
Protective Cover
Special Offers
  • Multiple Manufacturers
    Save up to 94% on Teaching Lab Essentials
    Promo Code25721
    Expires2025-05-01
    New Offer
    Save Now
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24156
    Expires2025-06-30
    Days
    59
    Hours
    9
    Minutes
    21
    Seconds
    45
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Product Title
IKA™ H 100 Silicone Protective Cover > 135deg.C