Learn More
Fisherbrand™ Silicone Pump Tubing
For use in field of medicine.
346.00 DKK - 1965.00 DKK
Specifications
Certifications/Compliance | Class 6 |
---|---|
Autoclavable | Autoclavable |
Material | Silicone |
Overall Length (Metric) | 1500 cm |
For Use With | Peristaltic Pump |
Product Code | Brand | Diameter (Metric) Inner | Thickness (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Thickness (Metric) | Price | Quantity & Availability | |||||
10141841
|
Fisherbrand™
FB56463 |
0.5 mm | 1.6 mm |
442.00 DKK
15m |
Estimated Shipment: 12-06-2025
Log in to see stock. |
|||||
10213321
|
Fisherbrand™
FB56465 |
0.8 mm | 1.6 mm |
346.00 DKK
15m |
Estimated Shipment: 17-06-2025
Log in to see stock. |
|||||
10631692
|
Fisherbrand™
FB56467 |
1.6 mm | 1.6 mm |
584.00 DKK
15m |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||||
10162031
|
Fisherbrand™
FB56487 |
1.6 mm | 2.4 mm |
788.00 DKK
15m |
Estimated Shipment: 12-06-2025
Log in to see stock. |
|||||
10283361
|
Fisherbrand™
FB56469 |
3.2 mm | 1.6 mm |
424.00 DKK
15m |
Estimated Shipment: 13-06-2025
Log in to see stock. |
|||||
10273411
|
Fisherbrand™
FB56489 |
3.2 mm | 2.4 mm |
861.00 DKK
15m |
Estimated Shipment: 02-06-2025
Log in to see stock. |
|||||
10172031
|
Fisherbrand™
FB56491 |
4.8 mm | 2.4 mm |
1115.00 DKK
15m |
Estimated Shipment: 12-06-2025
Log in to see stock. |
|||||
10335111
|
Fisherbrand™
FB56471 |
4.8 mm | 1.6 mm |
716.00 DKK
15m |
Estimated Shipment: 22-05-2025
Log in to see stock. |
|||||
10010061
|
Fisherbrand™
FB56493 |
6.4 mm | 2.4 mm |
1360.00 DKK
15m |
Estimated Shipment: 12-06-2025
Log in to see stock. |
|||||
10707501
|
Fisherbrand™
FB56473 |
6.4 mm | 1.6 mm |
708.00 DKK
15m |
Estimated Shipment: 12-06-2025
Log in to see stock. |
|||||
10599201
|
Fisherbrand™
FB56475 |
8 mm | 1.6 mm |
798.00 DKK
15m |
Estimated Shipment: 26-06-2025
Log in to see stock. |
|||||
10468582
|
Fisherbrand™
FB56495 |
8 mm | 2.4 mm |
1675.00 DKK
15m |
Estimated Shipment: 21-05-2025
Log in to see stock. |
|||||
10788061
|
Fisherbrand™
FB56497 |
9.6 mm | 2.4 mm |
1965.00 DKK
15m |
Estimated Shipment: 12-06-2025
Log in to see stock. |
|||||
Description
- FDA, NWC and BGA approved
- Certified to USP XXII class 6
- Autoclavable at 130°C in steam
- Sterilizable by gamma radiation without impairing quality
- Maybe opaque or translucent in appearance
Specifications
Class 6 | |
Silicone | |
Peristaltic Pump | |
Translucent |
Autoclavable | |
1500 cm | |
1 Each | |
Pump Tubing |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Silicone Pump Tubing