Learn More
Macherey-Nagel™ Silicone Standard Septa
Brand:  Macherey-Nagel™ 702612
 View more versions of this product
Product Code. 10176731
  • 388.00 DKK / Pack of 50
Estimated Shipment: 15-05-2025
to see stock.
Explore more special offers
Specifications
Standard Septum
60° Shore A
50 Pack
12 mm
Silicone
Product Title
Macherey-Nagel™ Silicone Standard Septa > O.D.: 12mm