Learn More
Macherey-Nagel™ Silicone Standard Septa
Brand:  Macherey-Nagel™ 702611
 View more versions of this product
Product Code. 10737452
  • 388.00 DKK / Pack of 50
Estimated Shipment: 20-05-2025
to see stock.
Explore more special offers
Specifications
Standard Septum
60° Shore A
50 Pack
11 mm
Silicone
Product Title
Macherey-Nagel™ Silicone Standard Septa > O.D.: 11mm