Learn More
Deutsch Neumann™ Silicone Rubber Stoppers
For safe and secure closure of laboratory apparatus. Deutsch Neumann™ Silicone Rubber Stoppers are translucent in color.
Brand: Deutsch Neumann™ 1010556
This item is not returnable.
View return policy
Description
- Acc. to DIN 12 871
- Durometer 50 Shore A
- Available in a wide range of sizes
Specifications
1/Ea. | |
Transparent | |
65 mm | |
Silicone Stopper |
200°C | |
56 mm | |
Silicone |