Learn More
Deutsch Neumann™ Silicone Rubber Stoppers
For safe and secure closure of laboratory apparatus. Deutsch Neumann™ Silicone Rubber Stoppers are translucent in color.
Brand: Deutsch Neumann™ 1010526+1001140
This item is not returnable.
View return policy
Description
- Acc. to DIN 12 871
- Durometer 50 Shore A
- Available in a wide range of sizes
Specifications
1/Ea. | |
26 mm | |
Silicone |
Transparent | |
32 mm | |
Silicone Stopper |