Learn More
Deutsch Neumann™ Silicone Rubber Stoppers
For safe and secure closure of laboratory apparatus. Deutsch Neumann™ Silicone Rubber Stoppers are translucent in color.
Brand:  Deutsch Neumann™ 1010526+1001140
Product Code. 10132864
  • 26.65 DKK / Each
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Acc. to DIN 12 871
  • Durometer 50 Shore A
  • Available in a wide range of sizes
Specifications
Specifications
1/Ea.
26 mm
Silicone
Transparent
32 mm
Silicone Stopper