Learn More
Bollé Safety Silium+ Platinum™ CSP Safety Glasses
Description
- Ultra-lightweight for extended wear
- Ultra lightweight frame
- Adjustable bridge
- Non-slip bridge
- Straight, highly flexible ergonomic temples
- PLATINUM coating
- All-round vision
Specifications
Specifications
Certifications/Compliance | Mechanical risk - Low energy impact - F; Mechanical risk- High speed particles at extreme temperatures - T; Risks relating to radiations - UV Protection - EN 170 - (2-1.2, 2C-1.2); Solar radiations risks: EN 172 |
Lens Coating | Anti-fog, Scratch-resistant |
Frame Color | Black |
Lens Material | Polycarbonate |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Videos
-
Bolle Safety Platinum efficiency demonstration
-
Bolle Safety Platinum coating
Documents
Product Information
Specification Sheets
SILIUMPLUS
File size: 1.1 MB
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
Bollé Safety Silium+ Platinum™ CSP Safety Glasses >
Spot an opportunity for improvement?Share a Content Correction