Learn More
Trajan Scientific and Medical™ SilTite™ Column Connectors
Ensures easy, leak-free installation for capillary columns without the use of any tools.
570.00 DKK - 1010.00 DKK
Specifications
For Use With (Equipment) Installation for Capillary Columns
View More Specs
Products 5
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
15258923
View Documents Promotion Details
Trajan Scientific and Medical™
073273
SilTite Kit 1/32 in. Column for Agilent
1010.00 DKK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 07-11-2024
to see stock.
Add to basket
15288913
View Documents Promotion Details
Trajan Scientific and Medical™
073223
SilTite Kit 1/16 in. Column for PerkinElmer
570.00 DKK
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 07-11-2024
to see stock.
Add to basket
15248923
View Documents Promotion Details
Trajan Scientific and Medical™
073233
SilTite Nuts for Shimadzu
875.00 DKK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 07-11-2024
to see stock.
Add to basket
15238923
View Documents Promotion Details
Trajan Scientific and Medical™
073230
SilTite Nuts for Thermo Scientific
875.00 DKK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 07-11-2024
to see stock.
Add to basket
15278913
View Documents Promotion Details
Trajan Scientific and Medical™
073219
SilTite Kit 1/32 in. Column for PerkinElmer
570.00 DKK
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 07-11-2024
to see stock.
Add to basket
Description
Description
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Specifications
Installation for Capillary Columns