Learn More
Trajan Scientific and Medical™ SilTite™ Column Connectors
Ensures easy, leak-free installation for capillary columns without the use of any tools.
570.00 DKK - 1010.00 DKK
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
15258923
|
Trajan Scientific and Medical™
073273 |
SilTite Kit 1/32 in. Column for Agilent |
1010.00 DKK
Each |
Estimated Shipment: 07-11-2024
Log in to see stock. |
|||||
15288913
|
Trajan Scientific and Medical™
073223 |
SilTite Kit 1/16 in. Column for PerkinElmer |
570.00 DKK
Pack of 10 |
Estimated Shipment: 07-11-2024
Log in to see stock. |
|||||
15248923
|
Trajan Scientific and Medical™
073233 |
SilTite Nuts for Shimadzu |
875.00 DKK
Pack of 5 |
Estimated Shipment: 07-11-2024
Log in to see stock. |
|||||
15238923
|
Trajan Scientific and Medical™
073230 |
SilTite Nuts for Thermo Scientific |
875.00 DKK
Pack of 5 |
Estimated Shipment: 07-11-2024
Log in to see stock. |
|||||
15278913
|
Trajan Scientific and Medical™
073219 |
SilTite Kit 1/32 in. Column for PerkinElmer |
570.00 DKK
Pack of 10 |
Estimated Shipment: 07-11-2024
Log in to see stock. |
|||||
Description
- Eliminates leaks
- No contamination
- No need to retighten after temperature cycling
- Cleaner spectra for better library matches
Specifications
Installation for Capillary Columns |