Learn More
Köttermann Single Bench Product Code.: 17332219
Köttermann Single Bench
Product Code. 17332219
600 x 900 x 900 mm, Trespa TopLab PLUS Worktop, Each
Click to view available options
Dimensions (D x W x H):
600 x 900 x 750 mm
600 x 900 x 900 mm
750 x 900 x 750 mm
750 x 900 x 900 mm
600 x 1200 x 750 mm
600 x 1200 x 900 mm
600 x 1500 x 750 mm
600 x 1500 x 900 mm
750 x 1200 x 750 mm
750 x 1200 x 900 mm
750 x 1500 x 750 mm
750 x 1500 x 900 mm
Top Style:
Melamine TopResist Worktop
Trespa TopLab PLUS Worktop
Unit Size:
Each
Product Code. 17332219
Brand: Köttermann 50500156
This item requires a price update prior to being ordered.
Request a Price for this item here.
This Single Bench fulfills a variety of tasks in the laboratory.
  • Available for sitting and standing work height
  • Stationary
Specifications
Dimensions (D x W x H) 600 x 900 x 900 mm
No. of Shelves 1
Top Style Trespa TopLab PLUS Worktop
Product Content Correction
Product Title
Köttermann Single Bench >