Learn More
Köttermann Single Bench
Click to view available options
Dimensions (D x W x H):
600 x 900 x 750 mm
600 x 900 x 900 mm
750 x 900 x 750 mm
750 x 900 x 900 mm
600 x 1200 x 750 mm
600 x 1200 x 900 mm
600 x 1500 x 750 mm
600 x 1500 x 900 mm
750 x 1200 x 750 mm
750 x 1200 x 900 mm
750 x 1500 x 750 mm
750 x 1500 x 900 mm
Top Style:
Melamine TopResist Worktop
Trespa TopLab PLUS Worktop
Unit Size:
Each
Description
- Available for sitting and standing work height
- Stationary
Specifications
Specifications
Dimensions (D x W x H) | 750 x 1500 x 750 mm |
No. of Shelves | 1 |
Top Style | Trespa TopLab PLUS Worktop |
Product Content Correction
Product Title
Köttermann Single Bench >
Spot an opportunity for improvement?Share a Content Correction