missing translation for 'onlineSavingsMsg'
Learn More

Sirtuin 1/SIRT1 Antibody (PCRP-SIRT1-1E11), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-14290IR

Product Code. 18310472

  • 4437.25 DKK / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Sirtuin 1/SIRT1 Monoclonal antibody specifically detects Sirtuin 1/SIRT1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Specifications

Specifications

Sirtuin 1/SIRT1
Monoclonal
DyLight 755
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human
Antibody
IgG1
Western Blot, Flow Cytometry, ELISA, Immunofluorescence
PCRP-SIRT1-1E11
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
EC 3.5.1, S. cerevisiae, homolog) 1, sirtuin 1
Recombinant full-length human Sirtuin 1/SIRT1protein (Uniprot: Q96EB6)
0.1 mL
Apoptosis, Chromatin Research, Cognition and Behavior, DNA Repair, Epigenetics
23411
Store at 4°C in the dark.
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers