Learn More
Notrax Skystep™ Ergonomic Rubber Mat with Moulded Edges
Ergonomically and economically engineered, the Skystep™ is designed to suction to the ground to prevent the mat from shifting while trapping air in its chambers. These air pockets combined with the resiliency of the rubber create an ultimate cushioning effect while keeping the mat lightweight.
392.00 DKK - 1040.00 DKK
Specifications
Color | Black |
---|---|
Material | Natural Rubber |
Type | Industrial and Commercial Matting |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
17792128
|
Notrax
455S0023BL |
90 cm |
392.00 DKK
Each |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||||
17702138
|
Notrax
455S0034BL |
120 cm |
792.00 DKK
Each |
Estimated Shipment: 01-08-2025
Log in to see stock. |
||||||
17712138
|
Notrax
455S0035BL |
150 cm |
1040.00 DKK
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
||||||
Description
- Ergonomically and economically engineered, the Skystep™ utilizes the power of air
- Skystep™ is designed to suction to the ground to prevent the mat from shifting while trapping air in its chambers
- These air pockets combined with the resiliency of the rubber create an ultimate cushioning effect while keeping the mat lightweight
- Integrated bevelled edges on all four sides to prevent tripping on the mat
- Loose lay with no installation required
- Slip resistance rating R9 according to DIN 51130 and BG-rule BGR181
- Free of DOP, free of DMF, free of ozone depleting substances, free of silicone and free of heavy metals
- 100% natural rubber compound
- General purpose applications
Specifications
Black | |
Natural Rubber | |
Industrial and Commercial Matting |