Learn More
Leica Microsystems Darkfield Slider
Improve unstained sample image clarity by increasing sample to background contrast via darkfield methodology. Leica Microsystems™ Darkfield Slider permits visualization of small structures typically overlooked by brightfield observation.
Brand:  Leica Microsystems 11501158
Product Code. 11954759
  • 415.00 DKK / Each
Estimated Shipment: 26-06-2025
to see stock.
Explore more special offers
Description
  • Improves clarity.
  • Increases contrast.
  • Fits DM model microscopes with slotted Abbe condensers.
Specifications
Specifications
DM Series microscopes with slotted Abbe condensers
Documents
One moment while we fetch your results.
Product Title
Leica Microsystems Darkfield Slider > For use with: DM Series microscopes with slotted Abbe condensers