Learn More
Hecht Karl™ Slides Product Code.: 11875930
Estimated Shipment: 25-09-2025
Only null left
Add to basket
Hecht Karl™ Slides
Product Code. 11875930
2, Pack of 50
Click to view available options
Well Count:
2
3
Unit Size:
Each
Pack of 50
Product Code. 11875930
Brand: Hecht Karl™ 42411010
Only null left
Estimated Shipment: 25-09-2025
Add to basket
Only null left
Add to basket
Perfectly suited for holding liquids and cultures for microscopic examination. Karl Hecht™ Cavity Slides/Plates are available with one or more cavities and in single slide or plate format.
  • Microscope slides/plates with one or more cavities.
  • Ground edges.
Specifications
Length (Metric) 76 mm
Width (Metric) 26 mm
Type Slide
Well Count 2
Quantity 1 Pack
Dimensions (L x W) 76 x 26 mm
Product Content Correction
Product Title
Hecht Karl™ Slides >