Learn More
Sludge Judge II Sampler System, 1.25" dia, 15 ft
Brand:  Cole Parmer 05465-30
Product Code. 15305512
  • 2695.00 DKK / Each
Estimated Shipment: 04-06-2025
to see stock.
Explore more special offers
Product Title
Sludge Judge II Sampler System, 1.25" dia, 15 ft >