Learn More
Moldex™ Smart FFP-Mask
Protects against Ozone, and nuisance relief
Brand:  Moldex™ 244501
 View more versions of this product
Product Code. 12392299
  • 1270.00 DKK / Pack of 10
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Description
Moldex Smart Special FFP2 NR D + Ozone 10 x WEL valved dust mask.
The activated layer of charcoal built into this mask offers relief from low levels of gases and vapours, often found in conjunction with particulate hazards. FFP2 NR D + Ozone 10 x WEL.
Specifications
Specifications
EN 149:2001+A1:2009
Smart FFP-Mask
Comfortable Nose Seal
Single-use
Nuisance Relief or Protection Against Ozone
10/Pk.