Learn More
Hecht Karl™ Soda Glass Assistant Test Tube
Brand: Hecht Karl™ 42769051
This item is not returnable.
View return policy
Specifications
Round bottom | |
No | |
Test Tube |
100Pack | |
16 mm |
Product Content Correction
Product Title
Hecht Karl™ Soda Glass Assistant Test Tube > Diameter: 16mm
Spot an opportunity for improvement?Share a Content Correction