Learn More
Gravis Glass Pill Bottles
Excellent for sampling and storage needs. Parisienne de Verrerie™ Glass Pill Bottles are manufactured from borosilicate glass.
Brand:  Gravis E00910M
Product Code. 11831623
  • 832.00 DKK / Pack of 100
Only null left
Estimated Shipment: 13-08-2025
Description
  • Can be closed with a snap cap
  • In acc. with US Pharmacopoeia Type I
Specifications
Specifications
Clear
8.5 mL
100
Polyethylene
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Gravis Glass Pill Bottles > Clear; Capacity: 8.5mL