missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-pentanesulfonate monohydrate, HPLC grade, Thermo Scientific Chemicals

Ion pair reagent

484.39 DKK - 4611.43 DKK

Chemical Identifiers

CAS 207605-40-1
Molecular Formula C5H13NaO4S
Molecular Weight (g/mol) 192.205
MDL Number MFCD00007541
InChI Key FPQYXAFKHLSWTI-UHFFFAOYSA-M
Synonym sodium pentane-1-sulfonate hydrate, 1-pentanesulfonic acid sodium salt monohydrate, sodium 1-pentanesulfonate monohydrate, potassium hydrate pentane-1-sulfonate, sodium 1-pentanesulfonate hydrate, sodium hydrate pentane-1-sulfonate, sodium pentane-1-sulfonate-water 1/1/1, 1-pentane sulphonic acid sodium salt monohydrate, 1-pentanesulfonic acid sodium salt monohydydrate, sodium 1-pentanesulfonate monohydrate, hplc grade
PubChem CID 23693099
IUPAC Name sodium;pentane-1-sulfonate;hydrate
SMILES CCCCCS(=O)(=O)[O-].O.[Na+]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11317138
SDS View Documents
Thermo Scientific Alfa Aesar
015714.06
5 g
484.39 DKK
5g
Please to purchase this item. Need a web account? Register with us today!
11307138
SDS View Documents
Thermo Scientific Alfa Aesar
015714.14
25 g
1334.69 DKK
25g
Please to purchase this item. Need a web account? Register with us today!
11397128
SDS View Documents
Thermo Scientific Alfa Aesar
015714.22
100 g
4611.43 DKK
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ion pair reagent.Sodium 1-pentanesulfonate monohydrate is used as a surfactant and a reversed-phase ion-pairing agent. It is utilized to study its effects on the properties of polyaniline, using Fourier transform infrared and UV-visible spectroscopy. Further, it is used for HPLC analysis of proteins and peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 245-208-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium 1-pentanesulfonate monohydrate, HPLC grade, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.