missing translation for 'onlineSavingsMsg'
Learn More
Sodium Carbonate Anhydrous, ACS Reagent Grade, Scharlab Product Code.: 11378534
compliance-icons

Sodium Carbonate Anhydrous, ACS Reagent Grade, Scharlab

Product Code. 11378534
500 g, 500g
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 11378534

Brand: Scharlab SO01160500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in analytical chemistry, as a laboratory reagent, to make sodium salts and as a detergent. Scharlab Sodium Carbonate Anhydrous, ACS Reagent Grade is used in glass manufacturing, the textile industry, photography, the pharmaceuticals industry and the food industry (E 500).

  • Synonyms: Anhydrous soda
  • Molecular Formula: NaCO3
  • Molar Mass: 105.99g/mol
  • CAS: 497-19-8
  • EIN ECS No.: 207-838-8

Specifications

Boiling Point 1600°C (decomposition)
Melting Point 854°C
CAS 497-19-8
Quantity 500 g
Molecular Formula CNa2O3
MDL Number MFCD00003494
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]C([O-])=O
IUPAC Name disodium carbonate
Molecular Weight (g/mol) 105.99
PubChem CID 10340
ChEBI CHEBI:29377
Formula Weight 105.99g/mol
Percent Purity ≥99.8% (acidimetric, on dried sample)
Grade ACS Reagent
Loss on Drying 1.0% max (300°C)
Show More Show Less
compliance-icons
Product Identifier
  • Sodium carbonate anhydrous, reagent grade, ACS, ISO, Reag. Ph Eur
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Carbonate Anhydrous, ACS Reagent Grade, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.