missing translation for 'onlineSavingsMsg'
Learn More

Sodium Cation Standard, 1mg/ml, Reagecon™

Product Code. 11596683 Shop All Reagecon Products
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in analytical chemistry and suitable for Ion Chromatography.

This standard is prepared, tested, certified and verified by following the exact same regime as already presented for ICP-MS Standards. It delivers confidence in the reliability of ion chromatography analysis. All are prepared from High Purity Starting materials and are traceable to N.I.S.T, where possible.

Specifications

Concentration 1mg/mL
Quantity 100 mL
Molecular Formula Na • H2O
Vapor Pressure 23hPa (at 20°C)
Analyte or Component Names Na+ ion in H2O matrix
Density 1.00296g/cm3
Melting Point 0°C
Boiling Point 100°C
Solubility Completely miscible
Physical Form Liquid
compliance-icons
Product Identifier
  • ION CHROMATOGRAPHY STANDARD SODIUM
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Supplemental information
  • MIXTURE LIST-Contains : sodium nitrate, containing in the dry statemore than 16,3 per cent by weight ofnitrogen
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.