Learn More
Sodium hydrogen sulfate, anhydrous, 90+%, remainder mainly sodium sulfate, Thermo Scientific Chemicals
Product Code. 15405095
Click to view available options
Quantity:
250 g
1000 g
5000 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7681-38-1
Molecular Formula HNaO4S
Molecular Weight (g/mol) 120.05
MDL Number MFCD00003531
InChI Key WBHQBSYUUJJSRZ-UHFFFAOYSA-M
Synonym sodium hydrogen sulfate, sodium bisulfate, sodium acid sulfate, bisulfate of soda, sodium hydrosulfate, fanal, sodium hydrogensulphate, niter cake, nitre cake, wc-perfect
PubChem CID 516919
SMILES [Na+].OS([O-])(=O)=O
Specifications
Melting Point 177°C to 180°C
pH 1.0
Quantity 5000 g
UN Number UN3260
Merck Index 14,8586
IUPAC Name sodium hydrogen sulfate
Formula Weight 120.06
Percent Purity ≥90%
Odor Odorless
Sensitivity Hygroscopic
Density 2.435
Assay remainder mainly sodium sulfate
Chemical Name or Material Sodium hydrogen sulfate, anhydrous