missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 0.1M/l Solution, Scharlab
Click to view available options
Quantity:
1 L
10 L
Unit Size:
1 L
10 L
Description
- Molecular Formula: NaOH
- Molar Mass: 40.00g/mol
- CAS: 1310-73-2
- EIN ECS No.: 215-185-5
- Factor: 0.999–1.001, uncertainty ±0.001
This volumetric solution was checked by means of potentiometric methods using Scharlau's potassium hydrogen phthalate volumetric standard. Scharlau's volumetric standards are directly traceable to the Standard Reference Materials from NIST (National Institute of Standards and Technology, USA).
Specifications
Specifications
Concentration | 0.1mol/L |
Concentration or Composition Notes | 1mL = 0.00400g NaOH |
Density | 1.00g/cm3 |
Quantity | 1 L |
Formula Weight | 40.00g/mol |
Normality | 0.1N |
Traceability to NIST | Traceable to NIST |
Physical Form | Solution |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Hydroxide, 0.1M/l Solution, Scharlab >
Spot an opportunity for improvement?Share a Content Correction