missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide Pellets, Pharmpur™, Scharlab
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
Description
- Synonyms: Caustic soda
- Molecular Formula: NaOH
- Molar Mass: 40.00g/mol
- CAS: 1310-73-2
- EIN ECS No.: 215-185-5
- Carbonates (as Na2CO3): max. 3%
- Insoluble Matter: Passes test
Specifications
Specifications
Color | Colorless |
Boiling Point | 1390°C |
CAS | 1310-73-2 |
Melting Point | 323°C |
Molecular Formula | HNaO |
Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
Solubility Information | Soluble in water at 20°C |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
Quantity | 1 kg |
SMILES | [OH-].[Na+] |
Show More |
Safety and Handling

Product Identifier
- Sodium hydroxide, pellets, extra pure, Pharmpur, Ph Eur, BP, NF
- Danger
- Skin corrosion/irritation Category 1 A
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Hydroxide Pellets, Pharmpur™, Scharlab >
Spot an opportunity for improvement?Share a Content Correction