missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide Solution Methanolic 1M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
659.00 DKK - 1590.00 DKK
Chemical Identifiers
CAS | 1310-73-2 |
---|---|
Molecular Formula | HNaO |
Molecular Weight (g/mol) | 39.997 |
MDL Number | 3548 |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
PubChem CID | 14798 |
ChEBI | CHEBI:32145 |
IUPAC Name | sodium;hydroxide |
SMILES | [OH-].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10765141
|
Fisher Chemical
J/7625/PB15 |
1 L | N/A | ||||||
10696832
|
Fisher Chemical
J/7625/17 |
2.5 L | N/A | ||||||
Description
Chemical Identifiers
1310-73-2 | |
39.997 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
14798 | |
sodium;hydroxide |
HNaO | |
3548 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
CHEBI:32145 | |
[OH-].[Na+] |
Specifications
Colorless | |
HNaO | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
Liquid |
1310-73-2 | |
3548 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
HDPE plastic bottle |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Sodium hydroxide, solution 1M, methanolic solution
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Methyl alcohol, Sodium hydroxide
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Hydroxide Solution Methanolic 1M (1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™