missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfate, Honeywell Fluka™

Product Code. 15993737
Click to view available options
Quantity:
500 g
2.5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

For pesticide residue analysis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-82-6
PubChem CID 24436
ChEBI CHEBI:32149

Specifications

Physical Form Solid
Quantity 2.5 kg
Grade Trace Analysis
Packaging Plastic Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.