Learn More
Memmert™ Software Atmocontrol, Base Licence For 1 Unit, Iq/Oq Doc. Available In De And En, For S750
Memmert™ AtmoCONTROL Software meets the requirements for the use of electronically stored data sets and electronic signatures.
Brand:  Memmert™ S750FDAQ1
Product Code. 16315977
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Software conforming to FDA AtmoCONTROL
  • Meets the requirements for the use of electronically stored data sets and electronic signatures as laid down in Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
  • Base licence for the control of one unit
  • Respective IQ/OQ documents available in German and English language (without surcharge)
Specifications
Specifications
Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
Models U, UF, TS, UNpa, S, I, ICP, IPP, IPS, HPP, ICH, 30, 55, 75, 110, 160, 260, 400, 450, 750, 106
Software
Videos
  • Memmert AtmoCONTROL (EN)
    Video Player is loading.
    Current Time 0:00
    Duration 1:36
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 1:36
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Memmert AtmoCONTROL (EN)
  • Memmert ControlCOCKPIT (EN)
    Video Player is loading.
    Current Time 0:00
    Duration 1:15
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 1:15
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Memmert ControlCOCKPIT (EN)
Product Title
Memmert™ Software Atmocontrol, Base Licence For 1 Unit, Iq/Oq Doc. Available In De And En, For S750 >