Learn More
Thermo Scientific™ SOLA™ SPE Cartridges
Achieve greater reproducibility with cleaner more consistent extracts compared to conventional SPE, phospholipid removal and protein precipitation products.
Brand:  Thermo Scientific™ 60109-004
1712.66 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15169653
  • 1721.22 DKK / Pack of 100
In Stock
Explore more special offers
Description
Grow your laboratory productivity. Thermo Scientific™ SOLA™ SPE Cartridges revolutionize solid phase extraction (SPE). This first fritless SPE product range enables greater sample throughput in bioanalytical laboratories where low elution volumes combined with greater reproducibility, lower failure rates and cleaner extracts are key to achieving better productivity.
SOLA products are available in a 10mg/1mL cartridge format. They provide highly reproducible, clean and consistent sample extracts for bioanalysis by utilizing the unique award winning fritless SPE technology. The enhanced performance of SOLA cartridges gives you greater confidence in your analytical results and lowers operating costs - without compromising ease of use or requiring complex method development.
SOLA cartridges provide unparalleled performance characteristics compared to conventional SPE, phospholipid removal and protein precipitation products.
What SOLA Cartridges Mean to Your Analysis
  • Higher levels of reproducibility
  • Higher levels of extract cleanliness
  • Reduced solvent requirements
  • Confidence in results
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 mL
10 mg
SOLA
SPE Cartridge
SOLA WCX SPE Cartridge
Bioanalytical Applications - Strong Ion-Exchange Retention of Strong Basic Compounds. Sorbent Charge Can be Activated or Deactivated. Complementary Reversed Phase Retention of Neutral Compounds.
Mixed Mode Weak Cation Exchanger, Polymeric
1 mL
Videos
  • Thermo Scientific - SOLA SPE cartridges and plates
    Thermo Scientific SOLA products revolutionise Solid Phase Extraction (SPE). This first fritless SPE product range provides greater reproducibility with cleaner, more consistent extracts.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Thermo Scientific SOLA SPE cartridges and plates
    • Thermo Scientific - SOLA SPE cartridges and plates
      Thermo Scientific SOLA products revolutionise Solid Phase Extraction (SPE). This first fritless SPE product range provides greater reproducibility with cleaner, more consistent extracts.
      Video Player is loading.
      Current Time 0:00
      Duration 2:19
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 2:19
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Thermo Scientific SOLA SPE cartridges and plates
    Product Title
    Thermo Scientific™ SOLA™ SPE Cartridges > 10mg/1mL; SOLA WCX SPE cartridge; 100 pack