missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18643-100UL
This item is not returnable.
View return policy
Description
SOX4 Polyclonal antibody specifically detects SOX4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
SOX4 | |
Polyclonal | |
Western Blot 1:4000, ELISA 1:100-1:1000, Immunohistochemistry 1:200 - 1:500, Immunoprecipitation 6 ul/IP, Immunohistochemistry-Paraffin 1:200-1:500 | |
Q06945 | |
Rabbit | |
Whole Antiserum | |
RUO | |
Primary | |
Human | |
Antisera |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Whole antiserum | |
ecotropic viral integration site 16, EVI16, SRY (sex determining region Y)-box 4, SRY-related HMG-box gene 4, transcription factor SOX-4 | |
Polyclonal antibody raised in rabbit against human SOX4 (SRY (sex determining region Y)-box 4), using two KLH-conjugated synthetic peptides containing a sequence from the N-terminus and from the C-terminus of the protein, respectively. | |
100 μg | |
Stem Cell Markers, Stem Cells | |
6659 | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |