Learn More
Lenz Laborglasintrumente™ Soxhlet Extractor Head
Extractor heads acc. to Soxhlet
Brand:  Lenz Laborglasintrumente™ 05400143
1079.00 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10284191
  • 1280.00 DKK / Each
Only null left
Estimated Shipment: 18-08-2025
Description
  • Extractor heads acc. to DIN 12602 and in addition to DIN standard
  • With cone NS 29/32
  • Includes glass NS stopcock
  • Made of DURAN™ tubing
Specifications
Specifications
150 mL
DURAN Tubing
Glass NS stopcock
NS 45/40
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lenz Laborglasintrumente™ Soxhlet Extractor Head > Capacity (Metric): 150 mL