Learn More
Bel-Art™ Roto-Flo™ Polystyrene Flow Indicator
Flow indicator with green paddle wheels go with the flow
Brand:  Bel-Art™ H19937-0003
Product Code. 11392214
  • 213.00 DKK / Each
Only null left
Estimated Shipment: 18-08-2025
Description
  • Fits 7.9 to 11mm (0.31 x 0.43 in.) I.D. tubing
  • Barbed tube ends
Specifications
Specifications
0/1 V
Measuring Tube
Flow Indicator
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Bel-Art™ Roto-Flo™ Polystyrene Flow Indicator > Polystyrene flow indicator; Green