Learn More
Merck Accessories for Swinnex Filter Holders
Accessories for filter holders for sample preparation.
Brand:  Merck SX00047RK
Product Code. 10229191
  • 4355.00 DKK / Each
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Description
MilliporeSigma Spare Parts, For Use With: For Swinnex 47, Includes: 10 Grid-together support and 10 boards for the base and 2 Plastic wrenches
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Not Autoclavable
For Swinnex 47
10 Grid-together support and 10 boards for the base and 2 Plastic wrenches
Spare Parts
Non-sterile
Laboratory Filter Holders or Cyclones