Learn More
Labcold™ Sparkfree Freezers
For the safe storage of potentially flammable items. Labcold™ Sparkfree Freezers come in both chest and upright models.
Brand:  Labcold™ RLVF1514
 View more versions of this product
Product Code. 15523851
Description
Description
  • CFC and HCFC free
  • Operator controlled defrost
  • Alarm/temperature display can be fitted as an optional extra
  • Chest freezers feature integral lid lock
  • Locks available for all upright freezers as an additional extra
Specifications
Specifications
400 L
Solid
RLVF1514
1855 mm
-12°C to -24°C
6 Adjustable
585 mm
600 mm
Product Title
Labcold™ Sparkfree Freezers > Capacity: 400L; Shelves: 6 adjustable; Temp.: -12° to -24°C; Style: Free standing