Learn More
Macherey-Nagel™ SPE Empty Column
Brand: Macherey-Nagel™ 730230
This item is not returnable.
View return policy
Specifications
15 mL | |
SPE | |
Empty Solid Phase Extraction Column |
Product Content Correction
Product Title
Macherey-Nagel™ SPE Empty Column > Column Capacity: 15mL
Spot an opportunity for improvement?Share a Content Correction