Learn More
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10003203
|
Invitrogen™
10668051 |
All Scales, 25 nmol |
32.51 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10377112
|
Invitrogen™
10629186 |
25 nmol |
1.45 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10424182
|
Invitrogen™
10629012 |
50 nmol |
1.85 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10022792
|
Invitrogen™
10629053 |
1 ÎĽmol |
17.32 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10357402
|
Invitrogen™
10915015 |
1 Vial, 1 ÎĽmol |
706.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10ÎĽmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Invitrogen™ Custom DNA Oligos Synthesis Services