Learn More
BINDER™ Specimen Temperature Display
Flexible Pt100 indoor temperature sensor
Brand:  BINDER™ 8012-2553
 View more versions of this product
Product Code. 30238364
  • 6764.67 DKK / Each
Only null left
Estimated Shipment: 23-06-2025
Description
Platinum 100 ohm temperature sensor for sample temperature display or control.
Specifications
Specifications
KB PRO 130, KB PRO 260, KB PRO 470, KB PRO 720, KB PRO 1060, KB PRO 1600
Steel
Temperature Sensor
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BINDER™ Specimen Temperature Display > For use with KB PRO 130, KB PRO 260, KB PRO 470, KB PRO 720, KB PRO 1060, KB PRO 1600