missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum Labs™ Spectra/Mesh™ Woven Stainless Steel Filter Disc
Hydrophilic stainless steel filter with very high strength for use in laboratory, industrial, or pharmaceutical applications.
Brand: Spectrum Labs™ 145825
1290.75 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Hydrophilic
- Good resistance against corrosives
- Resistant against pH 4 to 12
- Sterilize via irradiation/autoclave
Specifications
Stainless Steel | |
Laboratory, Industrial, Pharmaceutical | |
102 ÎĽm |
104 ÎĽm | |
45.2% | |
55 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spectrum Labs™ Spectra/Mesh™ Woven Stainless Steel Filter Disc > Pore Size: 104 μm
Spot an opportunity for improvement?Share a Content Correction