Learn More
Spectrum™ Spectra/Por™ Biotech Cellulose Ester (CE) Dialysis Membrane Tubing Product Code.: 11405959
Spectrum™ Spectra/Por™ Biotech Cellulose Ester (CE) Dialysis Membrane Tubing
Product Code. 11405959
10 mm, 25 kDa, Each
Click to view available options
Diameter (Metric):
6.4 mm
10 mm
15 mm
20 mm
24 mm
MWCO:
0.1 to 0.5 kDa
0.5 kDa
1 kDa
10 kDa
100 kDa
25 kDa
5 kDa
50 kDa
Unit Size:
10m
Each
Product Code. 11405959
Brand: Spectrum™ 131342
Selected MWCOs from 100 to 100,000 Daltons
  • CE tubing membranes are ideal for desalting, purification, and molecular separation
  • Supplied in 10m (33 ft.) lengths in two rolls of 5m each, in 0.1% sodium azide solution
Order Info
Note:Do not tie—must be used with Spectra/Por nylon closures (08-671-52, -53, -55). Strong polar solvents such as acetone, MEK, or dioxane will irreparably damage membranes. Contact Spectrum Technical Service, 1-800-634-3300, for more information.
Specifications
Product Type Spectra/Por Dialysis Membrane Tubing
MWCO 25 kDa
Material Cellulose Ester
Diameter (Metric) 10 mm
Diameter (English) 0.39 in.
Length (English) 33 ft.
Length (Metric) 10 m
Formulation Supplied in 0.1% sodium azide solution
Temperature Range (English) 39.2°F to 98.6°F
Temperature Range (Metric) 4°C to 37°C
Quantity 10 Pack
For Use With (Application) Ideal for desalting, purification, and molecular separation
Show More Show Less
Product Content Correction
Product Title
Spectrum™ Spectra/Por™ Biotech Cellulose Ester (CE) Dialysis Membrane Tubing >