Learn More
Cole-Parmer™ Replacement Sample Extraction Tool for Microvial Grinding Set
Sample Extraction Tool for removing ground samples from microvials which are used in the microvial set.
Brand: Cole-Parmer™ 6753C
This item is not returnable.
View return policy
Description
- Insert the tool into an individual vial, then push it through to clear sample from the tube.
Specifications
Replacement Extraction Tool | |
6757 Grinding Microvial Set (61043-06) | |
1 Ea. |